Skip to Content
Merck
All Photos(1)

Key Documents

59574

Supelco

α-Damascone

analytical standard

Synonym(s):

1-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-2-buten-1-one, 2,6,6-Trimethyl-1-crotonyl-2-cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
Beilstein:
2208705
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.490-1.500

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1=CCCC(C)(C)C1C(/C=C/C)=O

InChI

1S/C13H20O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5,7-8,12H,6,9H2,1-4H3/b7-5+

InChI key

CRIGTVCBMUKRSL-FNORWQNLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Damascone, a C13 norterpenoid is an important fragrance compound used in perfume industries due to its characteristic floral-fruity, rose-like odor. Commercially α-damascone is used in cosmetics, personal care products, detergents, etc.

Application

α-Damascone may be used as an analytical reference standard for the determination of the analyte in consumer products by gas chromatography-mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Principles and methodology for identification of fragrance allergens in consumer products
Gimenez-Arnau A, et al.
Contact Dermatitis, 47(6), 345-352 (2002)
Marcella Bovolenta et al.
The Journal of organic chemistry, 69(25), 8959-8962 (2004-12-04)
An efficient highly enantioselective (ee > or =99%) synthesis of alpha-ionone and alpha-damascone is described. Both enantiomers of title compounds were synthesized through two straightforward pathways diverging from enantiopure (R)- or (S)-alpha-cyclogeraniol. These versatile building blocks were obtained by regioselective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service