Skip to Content
Merck
All Photos(1)

Documents

32874

Supelco

Chlorimuron ethyl

PESTANAL®, analytical standard

Synonym(s):

Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15ClN4O6S
CAS Number:
Molecular Weight:
414.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(Cl)cc(OC)n2

InChI

1S/C15H15ClN4O6S/c1-3-26-13(21)9-6-4-5-7-10(9)27(23,24)20-15(22)19-14-17-11(16)8-12(18-14)25-2/h4-8H,3H2,1-2H3,(H2,17,18,19,20,22)

InChI key

NSWAMPCUPHPTTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Zhang et al.
International journal of molecular sciences, 20(12) (2019-06-27)
Klebsiella pneumoniae 2N3 is a strain of gram-negative bacteria that can degrade chlorimuron-ethyl and grow with chlorimuron-ethyl as the sole nitrogen source. The complete genome of Klebsiella pneumoniae 2N3 was sequenced using third generation high-throughput DNA sequencing technology. The genomic
Hao Zhang et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 45(6), 501-507 (2010-06-25)
Enrichment culturing of sludge taken from an industrial wastewater treatment pond led to the identification of a bacterium (Klebsiella jilinsis H. Zhang) that degrades chlorimuron-ethyl with high efficiency. Klebsiella jilinsis strain 2N3 grows with chlorimuron-ethyl as the sole nitrogen source
Xiaoli Zhang et al.
Environmental science and pollution research international, 18(3), 407-415 (2010-08-12)
Chlorimuron-ethyl has been widely used for the soybean production of China, but less information is available on the possible risk of long-term application of this herbicide. In this paper, soil samples were collected from the plots having been received 30 g
Myriam S Zawoznik et al.
Pest management science, 61(10), 1003-1008 (2005-05-28)
Possible side-effects of the acetolactate synthase (ALS)-inhibiting herbicide chlorimuron-ethyl on Bradyrhizobium japonicum (Kirchner & Jordan) in pure culture and on inoculated soybean plants growing under controlled conditions were investigated. Growth of B japonicum strain E109 was not affected by this
Jing Zhao et al.
Journal of agricultural and food chemistry, 54(14), 4948-4953 (2006-07-06)
Hybridomas secreting a monoclonal antibody (mAb) against the herbicide chlorimuron-ethyl (CE) were produced by fusing the mouse myeloma cell line (SP2/0) with splenocytes from a mouse immunized against the conjugate of the sulfonamide moiety of CE and bovine serum albumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service