Skip to Content
Merck
All Photos(1)

Documents

31720

Supelco

Prohexadione-Calcium

PESTANAL®, analytical standard

Synonym(s):

3,5-Dioxo-4-propionyl-cyclohexanecarboxylic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10CaO5
CAS Number:
Molecular Weight:
250.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Ca++].CC\C([O-])=C1/C(=O)CC(CC1=O)C([O-])=O

InChI

1S/C10H12O5.Ca/c1-2-6(11)9-7(12)3-5(10(14)15)4-8(9)13;/h5,11H,2-4H2,1H3,(H,14,15);/q;+2/p-2/b9-6-;

InChI key

NLKUPINTOLSSLD-BORNJIKYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Prohexadione-Calcium (Pro-Ca, prohexadione-Ca or BX-112) is a plant growth retardant which is used to inhibit gibberellin biosynthesis of plants. It generally reduces shoot elongation in plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of a new plant growth regulator prohexadione calcium (BX-112) on shoot elongation caused by exogenously applied gibberellins in rice (Oryza sativa L.) seedlings.
Nakayama, Ishizue, et al.
Plant & Cell Physiol., 31.2, 195-200 (1990)
Kai Jiang et al.
Plant physiology, 173(1), 825-835 (2016-12-03)
Gibberellin (GA) is a major plant hormone that regulates plant growth and development and is widely used as a plant growth regulator in agricultural production. There is an increasing demand for function-limited GA mimics due to the limitations on the
Shinsaku Ito et al.
Plant physiology, 174(2), 1250-1259 (2017-04-14)
Strigolactones (SLs) are a class of plant hormones that regulate diverse physiological processes, including shoot branching and root development. They also act as rhizosphere signaling molecules to stimulate the germination of root parasitic weeds and the branching of arbuscular mycorrhizal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service