Skip to Content
Merck
All Photos(3)

Documents

TZV000010

Millipore

Steridilutor® NEO Device

Devices without an expansion chamber for sample preparation and dilution. For liquids and soluble powders in vials.

Synonym(s):

Steridilutor® NEO device for sterility testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

Nylon 66 adapter (for needle)
PVC tubing
stainless steel (for needle)

Quality Level

Agency

EP (2.6.1)
JP (4.06)
USP 71

sterility

sterile; γ-irradiated

manufacturer/tradename

Steridilutor®

packaging

pkg of 10 blisters per box, Single packed

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)

shipped in

ambient

General description

The Steridilutor® NEO Device without expansion chamber is a tubing and needle assembly for difficult to dissolve powders, for dilution of super-potent antibiotics and pool products in vials. Pure products (solid or liquid) are difficult to rinse and residuals from products might be a major inhibitory factor in sterility testing. In the case of powders, a good dissolution is required to avoid any possible aggregate. A dilution step is strongly recommended for super potent antibiotics to reduce the concentration of the antimicrobial agent. This will reduce the possibility of binding of the agent to the filter. The small diameter double needle connects test products to the diluent. The diluted product is subsequently filtered with suitable Steritest® NEO devices.

Application

The Steridilutor® NEO Device without expansion chamber is used for difficult to dissolve powders, dilution of viscous products and pool products in vials. It helps for faster, easier and safer sterility testing.

Legal Information

STERIDILUTOR is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing is one of the most crucial steps in pharmaceutical product release. Regulatory-compliant membrane filtration sterility testing devices ensure the safety of pharmaceutical products.

Sterility testing is one of the most crucial steps in pharmaceutical product release. Regulatory-compliant membrane filtration sterility testing devices ensure the safety of pharmaceutical products.

Sterility testing is one of the most crucial steps in pharmaceutical product release. Regulatory-compliant membrane filtration sterility testing devices ensure the safety of pharmaceutical products.

Sterility testing is one of the most crucial steps in pharmaceutical product release. Regulatory-compliant membrane filtration sterility testing devices ensure the safety of pharmaceutical products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service