Skip to Content
Merck
All Photos(1)

Key Documents

SNAP2SH

Millipore

SNAP i.d. 2.0 IHC Slide Holders

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

manufacturer/tradename

SNAP id®

Quality Level

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry (frozen sections): suitable

compatibility

for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

storage temp.

room temp

General description

The SNAP i.d.® 2.0 System for IHC introduces a streamlined, medium through-put IHC capability to our SNAP i.d. 2.0 family. The frames and slide holders enable you to process up to 24 tissue slides, while our unique vacuum technology evenly removes your solutions - no more dipping or pouring.

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service