Skip to Content
Merck
All Photos(3)

Key Documents

F430-90

Phosphate Buffer

vial of 90 mL, pkg of 50 units, USP Butterfield′s Method, plastic vial (120ml flip top design), sterile; γ-irradiated by SER-TAIN process

Synonym(s):

Phosphate Buffer, Remel Phosphate Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

material

plastic vial (120ml flip top design)

Quality Level

Agency

USP Butterfield′s Method

sterility

sterile; γ-irradiated by SER-TAIN process

form

liquid

packaging

pkg of 50 units
vial of 90 mL

technique(s)

microbiological culture: suitable

pH

7.2±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phosphate buffer is a liquid medium designed for the qualitative detection of microorganisms in pharmaceutical and food samples. It serves as a sample diluent and as a medium for performing serial dilution. It prevents pH-associated variations while carrying out microbial experiments.

Application

Phosphate buffer is used for sample dilutions to detect microorganisms in pharmaceutical and food samples.

Legal Information

SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service