Skip to Content
Merck
All Photos(1)

Key Documents

BI0224

Sigma-Aldrich

Capping Reagent A 20% n-Methylimidazole in Acetonitrile

Biosynthesis Novabiochem®

Synonym(s):

dna

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

Quality Level

200
300

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

transition temp

flash point 5.5 °C

shipped in

ambient

storage temp.

2-30°C

Application

BI0224-0505 is packagedin GL45 glass bottles
BI0224-1005 is packagedin GL45 glass bottles
BI0224-P4000 is packaged in poly coated glass bottles
BI0224-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Assay (1-methylimidazole): 19.0-21.0% v/v
Assay (acetonitrile): 79.0-81.0% v/v
Color (APHA): 10 max
Component: N-Methylimidazole Purity: 99.0% min
Form: Clear liquid
Water (H2O): 200ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

41.9 °F - closed cup

Flash Point(C)

5.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service