Skip to Content
Merck
All Photos(1)

Key Documents

949159

Supelco

Filling nozzle (tap), self-closing, with G3/8" thread

for wall attachment

Synonym(s):

Filling nozzle wall

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121516
NACRES:
NA.03

shipped in

ambient

storage temp.

room temp

General description

Different environments and individual customer applications require special withdrawal options. Optimized nozzles are available for each special application, e.g. withdrawal inside a fume hood with a wall-attached filling nozzle while the barrel is installed inside a safety cabinet. This filling nozzle can be directly attached to the inner fume hood wall.
for wall attachment

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service