Skip to Content
Merck
All Photos(1)

Key Documents

8.55029

Sigma-Aldrich

N-Cyclohexylcarbodiimide,N′-methyl polystyrene

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carboxyl reactive

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

N-Cyclohexylcarbodiimide, N-methyl polystyrene [1] is an ideal tool for mediating solution phase coupling reactions. Typically, the amine and carboxylic acid are treated with a 2-3 fold excess of resin in DCM or DMF [2,3]; a slight excess of carboxyl component over amine component is often used to help drive the reaction to completion. As excess acid and urea by-products remain attached to the resin at the end of the reaction, the product can normally be isolated pure by filtration and evaporation of the reaction mixture. Reactions can be accelerated by the addition of auxiliary nucleophiles such as HOBt; these can be removed using polyamine- or TBD-polystyrene. This resin has also been used in conjunction with DMAP to mediate ester formation [4].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] G.M Weinshenker, et al. (1988) Org. Synth., Coll. Vol. VI, 951.
[2] Y. Guan, et al. (2000) J. Comb. Chem., 2, 297.
[3] K. Senton, et al. (2003) J. Comb. Chem., 5, 336.
[4] M. Cano, et al. (2002) J. Org. Chem., 67, 129.

Linkage

Replaces: 01-64-0211

Analysis Note

Color (visual): slightly beige to brown
Appearance of substance (visual): beads
Loading (determined by HPLC after reaction of Z-Phe-OH with cyclohexylamine): ≥ 1.3 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The polymer matrix is copoly (styrene-2 % DVB), 200 - 400 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service