Skip to Content
Merck
All Photos(2)

Key Documents

475984

Sigma-Aldrich

MyoVin-1

A pyrazolopyrimidine compound that blocks ADP release from the actomyosin complex and acts as a potent, reversible and uncompetitive inhibitor of actin-stimulated ATPase activity of myosin V.

Synonym(s):

MyoVin-1, Myosin V Inhibitor 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H26N6O3S
CAS Number:
Molecular Weight:
538.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(C1=CC=CC(SCC2=CC=CC(NC3=NC(NN=C4C5=CC=C(N6CCOCC6)C=C5)=C4C=N3)=C2)=C1)O

InChI key

JTMFLTNSCRPRHU-UHFFFAOYSA-N

General description

A pyrazolopyrimidine compound that blocks ADP release from the actomyosin complex and acts as a potent, reversible and uncompetitive inhibitor of actin-stimulated ATPase activity of myosin V (Ki = 6.3 and 5.5 µM by single- and double-headed myosin V). Displays excellent selectivity over myosin II and myosin IV (<10% inhibition at 50 µM) and only minimally inhibits the kinase activity of Cdk1/B, CHK1, Plk1, Abl, ERK2, CKII and Aurora-B (<10% inhibition at 100 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Islam, K., et al. 2010. Angew. Chem. Int. Ed.49, 8484.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Rita Costa et al.
eLife, 9 (2020-03-21)
Neurons have a membrane periodic skeleton (MPS) composed of actin rings interconnected by spectrin. Here, combining chemical and genetic gain- and loss-of-function assays, we show that in rat hippocampal neurons the MPS is an actomyosin network that controls axonal expansion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service