Skip to Content
Merck
All Photos(1)

Documents

1.30194

Millipore

HY-LiTE® Liquid Test Pens for Free ATP

Test pens for detection of Free ATP in liquid samples using HY-LiTE® 2/2A luminometers

Synonym(s):

Liquid ATP pens, Liquid ATP swabs, Liquid ATP tests, Liquid ATP samplers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.74

product line

HY-LiTE®

manufacturer/tradename

HY-LiTE®

packaging

pkg of (Pkg of 50 pens)

application(s)

cosmetics
food and beverages
water monitoring

compatibility

for use with HY-LITE® luminometer

detection method

bioluminescent

storage temp.

2-8°C

General description

The HY-LiTE® Liquid Test Pens for Free ATP are ready prepared Pens to measure the Free ATP (Adenosine triphosphate) in liquid samples using the HY-LiTE® 2/2A luminometer. ATP is detected by reaction with a luciferin/luciferase reagent in buffered solution.
Compared to the HY-LiTE® Liquid Test Pen for Total ATP (1.30102), the white sampling stick of the Liquid Pen for Free ATP does not contain a lysis reagent. Therefore, bacterial cells are not lysed and only the free ATP in the liquid solution is measured. A comparative measurement using both HY-LiTE® liquid pens (1.30102 Total ATP and 1.30194 Free ATP) can be performed in order to provide information on the proportion of cellularly bound ATP.

Application

HY-LiTE® Liquid Test Pens for Free ATP are used for monitoring CIP (Cleaning in Place) rinse waters, biomass monitoring in biocide water treatment, or other liquid samples by measuring with the HY-LiTE® 2/2A luminometer.

Suitability

For detection of Free ATP in liquid samples to monitor biomass

Linkage

Replaces: 1.30194.0022

Analysis Note

packaging: conforms
Appearance: Red caps
Performance test: conforms

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service