Skip to Content
Merck
All Photos(1)

Key Documents

1.10455

Millipore

Columbia agar (base)

for the cultivation of fastidious microorganisms (According harm. EP/USP/JP and ISO)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.3 (25 °C, 42 g/L in H2O)

solubility

42 g/L

bulk density

646 kg/m3

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Clostridium spp.

General description

for the cultivation of fastidious microorganisms (According harm. EP/USP/JP and ISO)

Analysis Note

Appearance (clearness)clear to slightly opalescentAppearance (color)yellowish-brownpH-value (25 °C)7.1 - 7.5Gel strength (Gelomat assay) (Base)≥ 50 gGel strength (Gelomat assay) (with blood)≥ 40 gTypical Composition (g/litre): Peptone from casein10.0; Peptone from meat 5.0; Heart extract 3.0; Extract from yeast 5.0; Starch 1.0; Sodium chloride 5.0; Agar-Agar 13.0.Growth promotion test in accordance with the harmonised method of EP, USP and JP.Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012))10 - 100Inoculum on reference medium (Staphylococcus aureus ATCC 6538 (WDCM 00032))10 - 100Inoculum on reference medium (Streptococcus pyogenes ATCC 12344)10 - 100Inoculum on reference medium (Streptococcus pyogenes ATCC 21059)10 - 100Inoculum on reference medium (Streptococcus pneumoniae ATCC 6301)10 - 100Inoculum on reference medium (Enterococcus faecalis ATCC 19433 (WDCM 00009))10 - 100Inoculum on reference medium (Bacillus cereus ATCC 11778 (WDCM 00001))10 - 100Inoculum on reference medium (Clostridium sporogenes ATCC 11437)10 - 100Inoculum on reference medium (Clostridium sporogenes ATCC 19404 (WDCM 00008))10 - 100Colony count (Escherichia coli ATCC 8739 (WDCM 00012))Colony count (Staphylococcus aureus ATCC 6538 (WDCM 00032))Colony count (Streptococcus pyogenes ATCC 12344)Colony count (Streptococcus pyogenes ATCC 21059)Colony count (Streptococcus pneumoniae ATCC 6301)Colony count (Enterococcus faecalis ATCC 19433 (WDCM 00009))Colony count (Bacillus cereus ATCC 11778 (WDCM 00001))Colony count (Clostridium sporogenes ATCC 11437)Colony count (Clostridium sporogenes ATCC 19404 (WDCM 00008))Recovery on test medium (Escherichia coli ATCC 8739 (WDCM 00012))≥ 70 %Recovery on test medium (Staphylococcus aureus ATCC 6538 (WDCM 00032))≥ 70 %Recovery on test medium (Streptococcus pyogenes ATCC 12344)≥ 70 %Recovery on test medium (Streptococcus pyogenes ATCC 21059)≥ 70 %Recovery on test medium (Streptococcus pneumoniae ATCC 6301)≥ 70 %Recovery on test medium (Enterococcus faecalis ATCC 19433 (WDCM 00009))≥ 70 %

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service