Skip to Content
Merck
All Photos(1)

Key Documents

P-114

Supelco

PEth 16:0/18:1 solution

1.0 mg/mL in methanol (as free phosphate), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H108NO8P
Molecular Weight:
942.42
UNSPSC Code:
41116107

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free phosphate)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−70°C

SMILES string

CCOP(OC[C@H](OC(CCCCCCCC=CCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C39H75O8P.C16H36N/c1-4-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-39(41)47-37(36-46-48(42,43)45-6-3)35-44-38(40)33-31-29-27-25-23-21-18-16-14-12-10-8-5-2;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h19-20,37H,4-18,21-36H2,1-3H3,(H,42,43);5-16H2,1-4H3/q;+1/p-1/b20-19-;/t37-;/m1./s1

InChI key

QDVSIAATLNPVDB-VYMFHCNISA-M

General description

PEth 16:0/18:1, also known as phosphatidylethanol 16:0/18:1, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/18:1 in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:1 levels is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:1 has also been investigated as a biomarker to confirm prenatal alcohol exposure (PAE).

Application


  • Alcohol consumption biomarker analysis: PEth 16:0/18:1 is used as a biomarker in whole blood and hair for monitoring alcohol use, proving essential in both clinical settings and research studies focused on substance abuse and its physiological impacts (Dumitrascu et al., 2024).

  • Diagnostic tool in traffic safety: Its application in dried blood samples from drivers involved in traffic accidents makes PEth 16:0/18:1 crucial for forensic science, aiding in the assessment of alcohol consumption as a factor in accidents (Daglioglu et al., 2022).

  • Method development in chromatography: PEth 16:0/18:1 is vital for developing methods to quantify phosphatidylethanol concentrations in novel sampling devices, enhancing the reliability of alcohol screening tests (Jett et al., 2023).

  • Advances in lipid chromatography: This compound is instrumental in refining chromatographic techniques to avoid the co-elution of phosphatidylethanols with unwanted phospholipids, improving the accuracy of lipidomics analyses (Maria et al., 2022).

  • Clinical biomarker validation: The validation of liquid chromatography tandem mass spectrometry (LC-MS/MS) methods for measuring erythrocyte PEth 16:0/18:1 emphasizes its role in clinical diagnostics, particularly in the accurate and sensitive detection of alcohol intake levels (White et al., 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service