Key Documents
SYNC154300
Synthware West condenser
joint: ST/NS 24/40, jacket L 300 mm
Synonym(s):
Synthware C154300, glass West condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C154300
jacket L
300 mm
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.