Key Documents
SLW1060/10D
Pyrex® Watch glasses
nominal diameter 80 mm
Synonym(s):
Pyrex, SciLabware 1060/10D
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
packaging
pkg of 10
manufacturer/tradename
1060/10D
SciLabware
nominal diameter
80 mm
General description
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Legal Information
PYREX is a registered trademark of Corning, Inc.