Skip to Content
Merck
All Photos(1)

Key Documents

923370

Sigma-Aldrich

6-((2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)oxy)pyrimidine-4-carboxylic acid

Synonym(s):

Rigid linker for PROTAC® development

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21N3O5
Molecular Weight:
335.36
UNSPSC Code:
12352106
NACRES:
NA.22

form

powder or crystals

Quality Level

reaction suitability

reagent type: linker

functional group

Boc
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(C1=CC(OC2CC3(C2)CN(C(OC(C)(C)C)=O)C3)=NC=N1)=O

Application

6-((2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)oxy)pyrimidine-4-carboxylic acid is useful as a rigid linker in PROTAC development for targeted protein degradation as well as chemical conjugates. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties. Developed in partnership with ComInnex.

Similar linkers in this series:
923311
923338
923419
923346
923427

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service