Skip to Content
Merck
All Photos(1)

Documents

88808

Sigma-Aldrich

(S)-(+)-1-Methoxy-2-propanol

≥98.5% (sum of enantiomers)

Synonym(s):

(S)-(+)-Propylene glycol 1-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2
CAS Number:
Molecular Weight:
90.12
Beilstein:
1718940
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (sum of enantiomers)

form

liquid

optical activity

[α]20/D +22±2°, c = 10% in chloroform

refractive index

n20/D 1.403

density

0.921 g/mL at 20 °C (lit.)

SMILES string

COC[C@H](C)O

InChI

1S/C4H10O2/c1-4(5)3-6-2/h4-5H,3H2,1-2H3/t4-/m0/s1

InChI key

ARXJGSRGQADJSQ-BYPYZUCNSA-N

Application

(S)-(+)-1-Methoxy-2-propanol can be used as a reactant to prepare:
  • (S)-1-methoxypropan-2-yl 4-methylbenzenesulfonate by reacting with tosyl chloride in the presence of pyridine.
  • Thiazolopyridine urea derivatives.
  • Quinazoline derivatives as potential inhibitors of EGFR/HER-2 tyrosine kinases.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of new quinazoline derivatives as irreversible dual EGFR/HER2 inhibitors and their anticancer activities-Part 1
D Debasis, et al.
Bioorganic & medicinal chemistry letters, 29(4), 591-596 (2019)
Stereocontrolled C (sp 3)-P bond formation with non-activated alkyl halides and tosylates
Yang C-T, et al.
Royal Society of Chemistry Advances, 7(40), 24652-24656 (2017)
Thiazolopyridine ureas as novel antitubercular agents acting through inhibition of DNA gyrase B
K Manoj G, et al.
Journal of Medicinal Chemistry, 56(21), 8834-8848 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service