Skip to Content
Merck
All Photos(1)

Key Documents

691623

Sigma-Aldrich

trans-Crotylboronic acid pinacol ester

95%

Synonym(s):

E-Crotylboronic acid pinacol ester, trans-2-(2-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19BO2
CAS Number:
Molecular Weight:
182.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.434

density

0.888 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C\C=C\CB1OC(C)(C)C(C)(C)O1

InChI

1S/C10H19BO2/c1-6-7-8-11-12-9(2,3)10(4,5)13-11/h6-7H,8H2,1-5H3/b7-6+

InChI key

ARSSMJZIAKUXEW-VOTSOKGWSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F

Flash Point(C)

63 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service