Skip to Content
Merck
All Photos(1)

Documents

44202

Sigma-Aldrich

Methyl dodecylbenzenesulfonate

≥95.0% (S)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32O3S
CAS Number:
Molecular Weight:
340.52
EC Number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (S)

form

liquid

refractive index

n20/D 1.496

density

1.027 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCc1ccc(cc1)S(=O)(=O)OC

InChI

1S/C19H32O3S/c1-3-4-5-6-7-8-9-10-11-12-15-18-16-13-14-17-19(18)23(20,21)22-2/h13-14,16-17H,3-12,15H2,1-2H3

InChI key

AIXYHMWIAPSTQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl dodecylbenzenesulfonate is an anionic surfactant.

Application

Methyl dodecylbenzenesulfonate may be employed as a reference standard for the determination of anionic surfactant by methylene blue active substance (MBAS) method.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of anionic surfactants in industrial effluents using a new highly sensitive surfactant-selective sensor.
Madunic-Cacic D, et al.
Sensor Letters, 7(1), 50-56 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service