Merck
  • Developmental YAPdeltaC determines adult pathology in a model of spinocerebellar ataxia type 1.
Developmental YAPdeltaC determines adult pathology in a model of spinocerebellar ataxia type 1.
Nature communications (2017-12-02)
Kyota Fujita, Ying Mao, Shigenori Uchida, Xigui Chen, Hiroki Shiwaku, Takuya Tamura, Hikaru Ito, Kei Watase, Hidenori Homma, Kazuhiko Tagawa, Marius Sudol, Hitoshi Okazawa
ABSTRACT
YAP and its neuronal isoform YAPdeltaC are implicated in various cellular functions. We found that expression of YAPdeltaC during development, but not adulthood, rescued neurodegeneration phenotypes of mutant ataxin-1 knock-in (Atxn1-KI) mice. YAP/YAPdeltaC interacted with RORα via the second WW domain and served as co-activators of its transcriptional activity. YAP/YAPdeltaC formed a transcriptional complex with RORα on cis-elements of target genes and regulated their expression. Both normal and mutant Atxn1 interacted with YAP/YAPdeltaC, but only mutant Atxn1 depleted YAP/YAPdeltaC from the RORα complex to suppress transcription on short timescales. Over longer periods, mutant Atxn1 also decreased RORα in vivo. Genetic supplementation of YAPdeltaC restored the RORα and YAP/YAPdeltaC levels, recovered YAP/YAPdeltaC in the RORα complex and normalized target gene transcription in Atxn1-KI mice in vivo. Collectively, our data suggest that functional impairment of YAP/YAPdeltaC by mutant Atxn1 during development determines the adult pathology of SCA1 by suppressing RORα-mediated transcription.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Calbindin-D-28K (EG-20) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Ataxin-1 Antibody, 11NQ, clone N76/8, clone N76/8, from mouse