Merck
Host-Mediated
The Plant cell (2017-11-01)
Tengfang Ling, Diana Bellin, Elodie Vandelle, Zahra Imanifard, Massimo Delledonne
ABSTRACT
Pathogens deliver effectors into plant cells to suppress immunity-related signaling. However, effector recognition by the host elicits a hypersensitive response (HR) that overcomes the inhibition of host signaling networks, restoring disease resistance. Signaling components are shared between the pathogen-associated molecular pattern-triggered immunity and effector-triggered immunity, and it is unclear how plants inactivate these effectors to execute the HR. Here, we report that, in
Product Description
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
3-Ethyl-3-(ethylaminoethyl)-1-hydroxy-2-oxo-1-triazene, ≥90%
Sigma-Aldrich
Monoclonal Anti-Biotin antibody produced in mouse, clone BN-34, ascites fluid
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution