Merck
  • Simultaneous quantification of cortisol and cortisone in urines from infants with packed-fiber solid-phase extraction coupled to HPLC-MS/MS.
Simultaneous quantification of cortisol and cortisone in urines from infants with packed-fiber solid-phase extraction coupled to HPLC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-07-25)
Chen Li, Zhao Zhang, Xiongwei Liu, Kangwei Shen, Pan Gu, Xuejun Kang
ABSTRACT
Cortisol and cortisone are two important glucocorticoids in human body, their interconversion is controlled by two isotypes of 11β-hydroxy steroid dehydrogenase (11β-HSD1 and 11β-HSD2). The ratio of urinary cortisol to cortisone can be used to assess the activity of 11β-HSDs. An analytical method to quantify urinary cortisol and cortisone using high performance liquid chromatographic tandem mass spectrometry following a packed-fiber solid-phase extraction (PFSPE) was developed. The proposed method was validated and applied to determine the urinary cortisol and cortisone concentrations in infants. Linearity was observed in the range of 0.6-150ng/mL for cortisol and 0.8-200ng/mL for cortisone. The intra-day RSD was 2.4-4.5% for cortisol and 3.3-6.2% for cortisone. Inter-day RSD was 3.7-6.6% for cortisol and 4.3-8.2% for cortisone. The recovery was 97.8±4.6% for cortisol and 98.9±4.4% for cortisone. The established method is simple and efficient for the quantification of urinary cortisol and cortisone and for indirectly assessing the activity of 11β-hydroxy steroid dehydrogenase.
Sigma-Aldrich
Cortisol-9,11,12,12-d4, ≥98 atom % D, ≥98% (CP)
Pricing and availability is not currently available.