Merck
  • Hierarchically self-assembled hexagonal honeycomb and kagome superlattices of binary 1D colloids.
Hierarchically self-assembled hexagonal honeycomb and kagome superlattices of binary 1D colloids.
Nature communications (2017-08-27)
Sung-Hwan Lim, Taehoon Lee, Younghoon Oh, Theyencheri Narayanan, Bong June Sung, Sung-Min Choi
ABSTRACT
Synthesis of binary nanoparticle superlattices has attracted attention for a broad spectrum of potential applications. However, this has remained challenging for one-dimensional nanoparticle systems. In this study, we investigate the packing behavior of one-dimensional nanoparticles of different diameters into a hexagonally packed cylindrical micellar system and demonstrate that binary one-dimensional nanoparticle superlattices of two different symmetries can be obtained by tuning particle diameter and mixing ratios. The hexagonal arrays of one-dimensional nanoparticles are embedded in the honeycomb lattices (for AB
Sigma-Aldrich
4-Vinylbenzoic acid, 97%
Sigma-Aldrich
Decyltrimethylammonium bromide, ≥98.0% (NT)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)