Merck
  • Stable engraftment of human microbiota into mice with a single oral gavage following antibiotic conditioning.
Stable engraftment of human microbiota into mice with a single oral gavage following antibiotic conditioning.
Microbiome (2017-08-02)
Christopher Staley, Thomas Kaiser, Lalit K Beura, Matthew J Hamilton, Alexa R Weingarden, Aleh Bobr, Johnthomas Kang, David Masopust, Michael J Sadowsky, Alexander Khoruts
ABSTRACT
Human microbiota-associated (HMA) animal models relying on germ-free recipient mice are being used to study the relationship between intestinal microbiota and human disease. However, transfer of microbiota into germ-free animals also triggers global developmental changes in the recipient intestine, which can mask disease-specific attributes of the donor material. Therefore, a simple model of replacing microbiota into a developmentally mature intestinal environment remains highly desirable. Here we report on the development of a sequential, three-course antibiotic conditioning regimen that allows sustained engraftment of intestinal microorganisms following a single oral gavage with human donor microbiota. SourceTracker, a Bayesian, OTU-based algorithm, indicated that 59.3 ± 3.0% of the fecal bacterial communities in treated mice were attributable to the donor source. This overall degree of microbiota engraftment was similar in mice conditioned with antibiotics and germ-free mice. Limited surveys of systemic and mucosal immune sites did not show evidence of immune activation following introduction of human microbiota. The antibiotic treatment protocol described here followed by a single gavage of human microbiota may provide a useful, complimentary HMA model to that established in germ-free facilities. The model has the potential for further in-depth translational investigations of microbiota in a variety of human disease states.
Sigma-Aldrich
Cefoperazone sodium salt, 870 - 1015 μg/mg anhydrous basis
Sigma-Aldrich
Ertapenem sodium, ≥90% (HPLC)