Merck
  • Evaluation of the performance of the IQ-Check kits and the USDA Microbiology Laboratory Guidebook methods for detection of Shiga toxin-producing Escherichia coli (STEC) and STEC and Salmonella simultaneously in ground beef.
Evaluation of the performance of the IQ-Check kits and the USDA Microbiology Laboratory Guidebook methods for detection of Shiga toxin-producing Escherichia coli (STEC) and STEC and Salmonella simultaneously in ground beef.
Journal of applied microbiology (2016-12-18)
G M Baranzoni, P M Fratamico, F Boccia, L K Bagi, G-H Kim, A Anastasio, T Pepe
ABSTRACT
To evaluate the performance of the IQ-Check kits and the USDA Microbiology Laboratory Guidebook (MLG) methods for detection of the top seven Shiga toxin-producing Escherichia coli (STEC) (O157:H7, O26, O45, O103, O111, O121 and O145) in ground beef and both STEC and Salmonella in co-inoculated samples. Ground beef samples inoculated with ~10 CFU of STEC or both STEC and Salmonella Typhimurium were stored at 4°C for 72 h, followed by screening with the IQ-Check and BAX System kit (MLG) methods that employ different enrichment media. STEC and S. Typhimurium were detected after 12 and 18 h and their presence was confirmed by colony isolation. Both methods were able to detect STEC in ground beef after 12 h of enrichment in samples inoculated with low levels of the pathogen. STEC and S. Typhimurium can be detected and isolated in co-inoculated ground beef samples. The IQ-Check methods are comparable to the MLG methods for detection of STEC and simultaneous detection of STEC and S. Typhimurium in seeded ground beef after a short enrichment time, thus the IQ-Check method can be useful for the food industry for rapid detection of these pathogens.
Product Description
Supelco
Cefixime trihydrate, suitable for microbiology, for ion-selective electrodes, ≥98.0% (HPLC)
Pricing and availability is not currently available.