Merck
Hijacking the E3 Ubiquitin Ligase Cereblon to Efficiently Target BRD4.
Chemistry & biology (2015-06-09)
Jing Lu, Yimin Qian, Martha Altieri, Hanqing Dong, Jing Wang, Kanak Raina, John Hines, James D Winkler, Andrew P Crew, Kevin Coleman, Craig M Crews
ABSTRACT
BRD4, a bromodomain and extraterminal domain (BET) family member, is an attractive target in multiple pathological settings, particularly cancer. While BRD4 inhibitors have shown some promise in MYC-driven malignancies such as Burkitt's lymphoma (BL), we show that BRD4 inhibitors lead to robust BRD4 protein accumulation, which may account for their limited suppression of MYC expression, modest antiproliferative activity, and lack of apoptotic induction. To address these limitations we designed ARV-825, a hetero-bifunctional PROTAC (Proteolysis Targeting Chimera) that recruits BRD4 to the E3 ubiquitin ligase cereblon, leading to fast, efficient, and prolonged degradation of BRD4 in all BL cell lines tested. Consequently, ARV-825 more effectively suppresses c-MYC levels and downstream signaling than small-molecule BRD4 inhibitors, resulting in more effective cell proliferation inhibition and apoptosis induction in BL. Our findings provide strong evidence that cereblon-based PROTACs provide a better and more efficient strategy in targeting BRD4 than traditional small-molecule inhibitors.
Sigma-Aldrich
N-Methylated pomalidomide, ≥98%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)