Merck
  • Microsomal membrane proteome of low grade diffuse astrocytomas: Differentially expressed proteins and candidate surveillance biomarkers.
Microsomal membrane proteome of low grade diffuse astrocytomas: Differentially expressed proteins and candidate surveillance biomarkers.
Scientific reports (2016-06-02)
Ravindra Varma Polisetty, Poonam Gautam, Manoj Kumar Gupta, Rakesh Sharma, Harsha Gowda, Durairaj Renu, Bhadravathi Marigowda Shivakumar, Akhila Lakshmikantha, Kiran Mariswamappa, Praveen Ankathi, Aniruddh K Purohit, Megha S Uppin, Challa Sundaram, Ravi Sirdeshmukh
ABSTRACT
Diffuse astrocytoma (DA; WHO grade II) is a low-grade, primary brain neoplasm with high potential of recurrence as higher grade malignant form. We have analyzed differentially expressed membrane proteins from these tumors, using high-resolution mass spectrometry. A total of 2803 proteins were identified, 340 of them differentially expressed with minimum of 2 fold change and based on ≥2 unique peptides. Bioinformatics analysis of this dataset also revealed important molecular networks and pathways relevant to tumorigenesis, mTOR signaling pathway being a major pathway identified. Comparison of 340 differentially expressed proteins with the transcript data from Grade II diffuse astrocytomas reported earlier, revealed about 190 of the proteins correlate in their trends in expression. Considering progressive and recurrent nature of these tumors, we have mapped the differentially expressed proteins for their secretory potential, integrated the resulting list with similar list of proteins from anaplastic astrocytoma (WHO Grade III) tumors and provide a panel of proteins along with their proteotypic peptides, as a resource that would be useful for investigation as circulatory plasma markers for post-treatment surveillance of DA patients.
Sigma-Aldrich
Anti-ENPP6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-HNRNPK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.