Merck
  • Insulin decreases atherosclerosis by inducing endothelin receptor B expression.
Insulin decreases atherosclerosis by inducing endothelin receptor B expression.
JCI insight (2016-05-21)
Kyoungmin Park, Akira Mima, Qian Li, Christian Rask-Madsen, Pingnian He, Koji Mizutani, Sayaka Katagiri, Yasutaka Maeda, I-Hsien Wu, Mogher Khamaisi, Simone Rordam Preil, Ernesto Maddaloni, Ditte Sørensen, Lars Melholt Rasmussen, Paul L Huang, George L King
ABSTRACT
Endothelial cell (EC) insulin resistance and dysfunction, caused by diabetes, accelerates atherosclerosis. It is unknown whether specifically enhancing EC-targeted insulin action can decrease atherosclerosis in diabetes. Accordingly, overexpressing insulin receptor substrate-1 (IRS1) in the endothelia of Apoe-/- mice (Irs1/Apoe-/-) increased insulin signaling and function in the aorta. Atherosclerosis was significantly reduced in Irs1/ApoE-/- mice on diet-induced hyperinsulinemia and hyperglycemia. The mechanism of insulin's enhanced antiatherogenic actions in EC was related to remarkable induction of NO action, which increases endothelin receptor B (EDNRB) expression and intracellular [Ca2+]. Using the mice with knockin mutation of eNOS, which had Ser1176 mutated to alanine (AKI), deleting the only known mechanism for insulin to activate eNOS/NO pathway, we observed that IRS1 overexpression in the endothelia of Aki/ApoE-/- mice significantly decreased atherosclerosis. Interestingly, endothelial EDNRB expression was selectively reduced in intima of arteries from diabetic patients and rodents. However, endothelial EDNRB expression was upregulated by insulin via P13K/Akt pathway. Finally EDNRB deletion in EC of Ldlr-/- and Irs1/Ldlr-/- mice decreased NO production and accelerated atherosclerosis, compared with Ldlr-/- mice. Accelerated atherosclerosis in diabetes may be reduced by improving insulin signaling selectively via IRS1/Akt in the EC by inducing EDNRB expression and NO production.
Sigma-Aldrich
Anti-Galectin-3 Antibody, clone M3/38, clone M3/38, from rat
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-phospho-IRS1 (Tyr608) mouse/ (Tyr612) human Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.