Merck
Liquid Structure of CO
The journal of physical chemistry. B (2016-10-27)
Quintin R Sheridan, Seungmin Oh, Oscar Morales-Collazo, Edward W Castner, Joan F Brennecke, Edward J Maginn
ABSTRACT
A combination of X-ray scattering experiments and molecular dynamics simulations were conducted to investigate the structure of ionic liquids (ILs) which chemically bind CO
Sigma-Aldrich
Pyrrole-2-carbonitrile, 96%
Sigma-Aldrich
Triethylphosphine, 99%
Sigma-Aldrich
1-Bromooctane, 99%