Merck
  • Decreased demand for olfactory periglomerular cells impacts on neural precursor cell viability in the rostral migratory stream.
Decreased demand for olfactory periglomerular cells impacts on neural precursor cell viability in the rostral migratory stream.
Scientific reports (2016-08-31)
Anika Langenfurth, Song Gu, Verena Bautze, Caiyi Zhang, Julia E Neumann, Ulrich Schüller, Kristin Stock, Susanne A Wolf, Anna-Maria Maier, Giorgia Mastrella, Andrew Pak, Hongwei Cheng, Roland E Kälin, Kenn Holmbeck, Jörg Strotmann, Helmut Kettenmann, Rainer Glass
ABSTRACT
The subventricular zone (SVZ) provides a constant supply of new neurons to the olfactory bulb (OB). Different studies have investigated the role of olfactory sensory input to neural precursor cell (NPC) turnover in the SVZ but it was not addressed if a reduced demand specifically for periglomerular neurons impacts on NPC-traits in the rostral migratory stream (RMS). We here report that membrane type-1 matrix metalloproteinase (MT1-MMP) deficient mice have reduced complexity of the nasal turbinates, decreased sensory innervation of the OB, reduced numbers of olfactory glomeruli and reduced OB-size without alterations in SVZ neurogenesis. Large parts of the RMS were fully preserved in MT1-MMP-deficient mice, but we detected an increase in cell death-levels and a decrease in SVZ-derived neuroblasts in the distal RMS, as compared to controls. BrdU-tracking experiments showed that homing of NPCs specifically to the glomerular layer was reduced in MT1-MMP-deficient mice in contrast to controls while numbers of tracked cells remained equal in other OB-layers throughout all experimental groups. Altogether, our data show the demand for olfactory interneurons in the glomerular layer modulates cell turnover in the RMS, but has no impact on subventricular neurogenesis.
Sigma-Aldrich
BrdU Cell Proliferation Assay, This proliferation assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells.
Pricing and availability is not currently available.