Merck
  • Differentiation of Spiral Ganglion-Derived Neural Stem Cells into Functional Synaptogenetic Neurons.
Differentiation of Spiral Ganglion-Derived Neural Stem Cells into Functional Synaptogenetic Neurons.
Stem cells and development (2016-03-30)
Xiaoyang Li, Alicia Aleardi, Jue Wang, Yang Zhou, Rodrigo Andrade, Zhengqing Hu
ABSTRACT
Spiral ganglion neurons (SGNs) are usually damaged in sensorineural hearing loss. SGN-derived neural stem cells (NSCs) have been identified and proposed to differentiate into neurons to replace damaged SGNs. However, it remains obscure whether SGN-NSC-derived neurons (ScNs) are electrophysiologically functional and possess the capability to form neural connections. Here, we found that SGN-derived cells demonstrated NSC characteristics and differentiated into SGN-like glutamatergic neurons. Neurotrophins significantly increased neuronal differentiation and neurite length of ScNs. Patch clamp recording revealed that ScNs possessed SGN-like NaV and HCN channels, suggesting electrophysiological function. FM1-43 staining and synaptic protein immunofluorescence showed ScNs possess the ability to form neural connections. Astrocyte-conditioned medium was able to stimulate ScNs to express synaptic proteins. These data suggested that neurotrophins are able to stimulate postnatal SGN-NSCs to differentiate into functional glutamatergic ScNs with the capability to form synaptic connections in vitro.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-ATH 1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, clone 3C10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken