Merck
  • A Sensitive IHC Method for Monitoring Autophagy-Specific Markers in Human Tumor Xenografts.
A Sensitive IHC Method for Monitoring Autophagy-Specific Markers in Human Tumor Xenografts.
Journal of biomarkers (2016-06-02)
Helen He, Yu Yang, Zhongmin Xiang, Lunyin Yu, Jouhara Chouitar, Jie Yu, Natalie Roy D'Amore, Ping Li, Zhi Li, Douglas Bowman, Matthew Theisen, James E Brownell, Stephen Tirrell
ABSTRACT
Objective. Use of tyramide signal amplification (TSA) to detect autophagy biomarkers in formalin fixed and paraffin embedded (FFPE) xenograft tissue. Materials and Methods. Autophagy marker regulation was studied in xenograft tissues using Amp HQ IHC and standard IHC methods. Results. The data demonstrate the feasibility of using high sensitivity TSA IHC assays to measure low abundant autophagy markers in FFPE xenograft tissue.
Sigma-Aldrich
MISSION® Lentiviral Packaging Mix, Production of replication incompetent viral particles
Pricing and availability is not currently available.