Merck
  • Deletion of tumor necrosis factor death receptor inhibits amyloid beta generation and prevents learning and memory deficits in Alzheimer's mice.
Deletion of tumor necrosis factor death receptor inhibits amyloid beta generation and prevents learning and memory deficits in Alzheimer's mice.
The Journal of cell biology (2007-08-29)
Ping He, Zhenyu Zhong, Kristina Lindholm, Lilian Berning, Wendy Lee, Cynthia Lemere, Matthias Staufenbiel, Rena Li, Yong Shen
ABSTRACT
The tumor necrosis factor type 1 death receptor (TNFR1) contributes to apoptosis. TNFR1, a subgroup of the TNFR superfamily, contains a cytoplasmic death domain. We recently demonstrated that the TNFR1 cascade is required for amyloid beta protein (Abeta)-induced neuronal death. However, the function of TNFR1 in Abeta plaque pathology and amyloid precursor protein (APP) processing in Alzheimer's disease (AD) remains unclear. We report that the deletion of the TNFR1 gene in APP23 transgenic mice (APP23/TNFR1(-/-)) inhibits Abeta generation and diminishes Abeta plaque formation in the brain. Genetic deletion of TNFR1 leads to reduced beta-secretase 1 (BACE1) levels and activity. TNFR1 regulates BACE1 promoter activity via the nuclear factor-kappaB pathway, and the deletion of TNFR1 in APP23 transgenic mice prevents learning and memory deficits. These findings suggest that TNFR1 not only contributes to neurodegeneration but also that it is involved in APP processing and Abeta plaque formation. Thus, TNFR1 is a novel therapeutic target for AD.
Sigma-Aldrich
1-Ethyl-2-pyrrolidone, 98%