Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Pricing and availability is not currently available.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Pricing and availability is not currently available.
Sigma-Aldrich
Carestream® Processing chemicals
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max™, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec™, reagent grade, 99%
Pricing and availability is not currently available.