Merck
  • High Aspect Ratio Carbon Nanotube Membranes Decorated with Pt Nanoparticle Urchins for Micro Underwater Vehicle Propulsion via H2O2 Decomposition.
High Aspect Ratio Carbon Nanotube Membranes Decorated with Pt Nanoparticle Urchins for Micro Underwater Vehicle Propulsion via H2O2 Decomposition.
ACS nano (2015-06-25)
Kevin M Marr, Bolin Chen, Eric J Mootz, Jason Geder, Marius Pruessner, Brian J Melde, Richard R Vanfleet, Igor L Medintz, Brian D Iverson, Jonathan C Claussen
ABSTRACT
The utility of unmanned micro underwater vehicles (MUVs) is paramount for exploring confined spaces, but their spatial agility is often impaired when maneuvers require burst-propulsion. Herein we develop high-aspect ratio (150:1), multiwalled carbon nanotube microarray membranes (CNT-MMs) for propulsive, MUV thrust generation by the decomposition of hydrogen peroxide (H2O2). The CNT-MMs are grown via chemical vapor deposition with diamond shaped pores (nominal diagonal dimensions of 4.5 × 9.0 μm) and subsequently decorated with urchin-like, platinum (Pt) nanoparticles via a facile, electroless, chemical deposition process. The Pt-CNT-MMs display robust, high catalytic ability with an effective activation energy of 26.96 kJ mol(-1) capable of producing a thrust of 0.209 ± 0.049 N from 50% [w/w] H2O2 decomposition within a compact reaction chamber of eight Pt-CNT-MMs in series.
Sigma-Aldrich
Chloroplatinic acid hexahydrate, BioXtra
Sigma-Aldrich
Chloroplatinic acid hexahydrate, ACS reagent, ≥37.50% Pt basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%