Merck
  • Serotonergic, brain volume and attentional correlates of trait anxiety in primates.
Serotonergic, brain volume and attentional correlates of trait anxiety in primates.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2015-01-15)
Yevheniia Mikheenko, Yoshiro Shiba, Stephen Sawiak, Katrin Braesicke, Gemma Cockcroft, Hannah Clarke, Angela C Roberts
ABSTRACT
Trait anxiety is a risk factor for the development and maintenance of affective disorders, and insights into the underlying brain mechanisms are vital for improving treatment and prevention strategies. Translational studies in non-human primates, where targeted neurochemical and genetic manipulations can be made, are critical in view of their close neuroanatomical similarity to humans in brain regions implicated in trait anxiety. Thus, we characterised the serotonergic and regional brain volume correlates of trait-like anxiety in the marmoset monkey. Low- and high-anxious animals were identified by behavioral responses to a human intruder (HI) that are known to be sensitive to anxiolytic drug treatment. Extracellular serotonin levels within the amygdala were measured with in vivo microdialysis, at baseline and in response to challenge with the selective serotonin reuptake inhibitor, citalopram. Regional brain volume was assessed by structural magnetic resonance imaging. Anxious individuals showed persistent, long-term fearful responses to both a HI and a model snake, alongside sustained attention (vigilance) to novel cues in a context associated with unpredictable threat. Neurally, high-anxious marmosets showed reduced amygdala serotonin levels, and smaller volumes in a closely connected prefrontal region, the dorsal anterior cingulate cortex. These findings highlight behavioral and neural similarities between trait-like anxiety in marmosets and humans, and set the stage for further investigation of the processes contributing to vulnerability and resilience to affective disorders.
Product Description
USP
Citalopram hydrobromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citalopram hydrobromide, ≥98% (HPLC)
Citalopram hydrobromide, European Pharmacopoeia (EP) Reference Standard
Citalopram for system suitability, European Pharmacopoeia (EP) Reference Standard