Merck
  • Thin-layer chromatography/direct analysis in real time time-of-flight mass spectrometry and isotope dilution to analyze organophosphorus insecticides in fatty foods.
Thin-layer chromatography/direct analysis in real time time-of-flight mass spectrometry and isotope dilution to analyze organophosphorus insecticides in fatty foods.
Journal of chromatography. A (2014-12-03)
Osamu Kiguchi, Kazuko Oka, Masafumi Tamada, Takashi Kobayashi, Jun Onodera
ABSTRACT
To assess food safety emergencies caused by highly hazardous chemical-tainted foods, simultaneous analysis of organophosphorus insecticides in fatty foods such as precooked foods was conducted using thin-layer chromatography/direct analysis in real time time-of-flight mass spectrometry (TLC/DART-TOFMS) and isotope dilution technique. Polar (methamidophos and acephate) and nonpolar organophosphorus insecticides (fenitrothion, diazinon, and EPN) were studied. Experiments to ascertain chromatographic patterns using TLC/DART-TOFMS reveal that it was more useful than GC/MS or GC/MS/MS for the simultaneous analyses of polar and nonpolar pesticides, while obviating the addition of a protective agent for tailing effects of polar pesticides. Lower helium gas temperature (260°C) for DART-TOFMS was suitable for the simultaneous analysis of target pesticides. Linearities were achieved respectively at a lower standard concentration range (0.05-5 μg) for diazinon and EPN and at a higher standard concentration range (2.5-25 μg) for methamidophos, acephate, and fenitrothion. Their respective coefficients of determination were ≥ 0.9989 and ≥ 0.9959. A few higher repeatabilities (RSDs) for diazinon and EPN were found (>20%), although isotope dilution technique was used. Application to the HPTLC plate without an automatic TLC sampler might be inferred as a cause of their higher RSDs. Detection limits were estimated in the higher picogram range for diazinon and EPN, and in the lower nanogram range for methamidophos, acephate, and fenitrothion. Aside from methamidophos, recovery results (n=3) obtained using a highly insecticide-tainted fatty food (dumpling) and raw food (grapefruit) samples (10mg/kg) using TLC/DART-TOFMS with both complex and simpler cleanups were not as susceptible to matrix effects (95-121%; RSD, 1.3-14%) as those using GC/MS/MS (102-117%; RSD, 0.4-8.5%), although dumpling samples using GC/MS were remarkably susceptible to matrix effects. The coupled method of TLC with simpler cleanup and DART-TOFMS can be regarded as the same analytical tool as GC/MS/MS, which is useful to assess food safety emergencies caused by highly hazardous chemical-tainted foods.
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Cyclohexane, JIS special grade
Sigma-Aldrich
Cyclohexane, suitable for HPLC
Sigma-Aldrich
Cyclohexane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Fenitrothion, PESTANAL®, analytical standard
Sigma-Aldrich
Chrysene-d12, 98 atom % D
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%