Merck
  • Fluorescence derivatization method for sensitive chromatographic determination of zidovudine based on the Huisgen reaction.
Fluorescence derivatization method for sensitive chromatographic determination of zidovudine based on the Huisgen reaction.
Journal of chromatography. A (2014-07-09)
Yuki Maeda, Naoya Kishikawa, Kaname Ohyama, Mitsuhiro Wada, Rie Ikeda, Naotaka Kuroda
ABSTRACT
A novel pre-column fluorescence derivatization method for chromatographic analysis of azide compounds was developed based on the Huisgen reaction, which is a specific cycloaddition reaction between an alkyne and an azide. We designed and synthesized a fluorescent alkyne, 2-(4-ethynylphenyl)-4,5-diphenyl-1H-imidazole (DIB-ET) as a reagent. DIB-ET has a lophine skeleton carrying an alkyne acting as fluorophore and reactive center, respectively. In order to evaluate the practicality of DIB-ET, a high-performance liquid chromatography with fluorescence detection method was developed for the determination of zidovudine as a model of azide compound. Zidovudine could be reacted with DIB-ET in the presence of copper(II) sulfate and L-ascorbic acid as catalysts, and the formed derivative was detected fluorometrically. The proposed method allows sensitive and selective determination of zidovudine in plasma samples with the detection limit of 0.28ngmL(-1) at a S/N=3. Finally, the proposed method could be applied to determine the zidovudine concentration in rat plasma after administration of zidovudine without interference from biological components.
Product Description
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Ethynylbenzaldehyde, 97%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v)
Sigma-Aldrich
Copper(II) sulfate, ≥97.5%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, reagent grade