Merck
  • Polysulfide promotes neuroblastoma cell differentiation by accelerating calcium influx.
Polysulfide promotes neuroblastoma cell differentiation by accelerating calcium influx.
Biochemical and biophysical research communications (2015-03-10)
Shin Koike, Norihiro Shibuya, Hideo Kimura, Kazuyuki Ishii, Yuki Ogasawara
ABSTRACT
Polysulfides are a typical type of bound sulfur, which is physiologically stable form of sulfur species, derived from the hydrogen sulfide (H2S) that is generated endogenously in cells. We previously reported that bound sulfur protects neuronal cells from oxidative injury. In the present study, we demonstrated that polysulfides inhibited cell growth and promoted neurite outgrowth in mouse neuroblastoma Neuro2A (N2A) cells. However, Na2S showed no effect on neurite outgrowth in N2A cells. Furthermore, 2-APB and SKF96365, which are typical transient receptor potential (TRP) channel inhibitors, suppressed the neurite outgrowth induced by Na2S4. These new findings suggest that bound sulfur could induce neurite outgrowth and cell differentiation of N2A cells by accelerating calcium influx.
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium sulfide nonahydrate, ≥99.99% trace metals basis
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF
Sigma-Aldrich
Sodium sulfide nonahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium sulfide nonahydrate, ≥98.0%