Merck
  • Substrate- and dose-dependent drug interactions with grapefruit juice caused by multiple binding sites on OATP2B1.
Substrate- and dose-dependent drug interactions with grapefruit juice caused by multiple binding sites on OATP2B1.
Pharmaceutical research (2014-02-20)
Yoshiyuki Shirasaka, Takanori Mori, Yukiko Murata, Takeo Nakanishi, Ikumi Tamai
ABSTRACT
OATP2B1-mediated grapefruit juice (GFJ)-drug interactions are substrate-dependent; for example, GFJ ingestion significantly reduces bioavailability of fexofenadine, but not pravastatin. In the present study, we aimed to establish whether this observation can be explained by the presence of multiple binding sites (MBS) on OATP2B1. OATP2B1-mediated drug uptake was evaluated using a Xenopus oocyte expression system. Drug concentration was quantified by LC/MS/MS analysis. OATP2B1-mediated uptake of pravastatin and fexofenadine exhibited biphasic saturation kinetics, indicating the presence of MBS on OATP2B1. GFJ strongly inhibited pravastatin uptake mediated by the high-affinity site on OATP2B1, while no significant inhibition of the low-affinity site was observed. In contrast, high-affinity transport of fexofenadine was only modestly inhibited by GFJ, while significant inhibition of the low-affinity site was observed. Contribution analysis indicated that both drugs are transported via the low-affinity site on OATP2B1 at therapeutically relevant concentrations. These findings indicate that only fexofenadine is expected to interact with GFJ on OATP2B1 at therapeutic concentrations, in accordance with the clinical observations. Substrate- and dose-dependent GFJ-drug interactions mediated by OATP2B1 might be explained in terms of the presence of MBS: interaction occurs only when drug and GFJ components share the same binding site on OATP2B1.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 1 M
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%