Merck
  • Semaphorin 4D expression is associated with a poor clinical outcome in cervical cancer patients.
Semaphorin 4D expression is associated with a poor clinical outcome in cervical cancer patients.
Microvascular research (2014-03-08)
Huidong Liu, Yanmei Yang, Jianbing Xiao, Shucai Yang, Yan Liu, Wenhui Kang, Xinlei Li, Fengmin Zhang
ABSTRACT
Lymphangiogenesis is thought to be essential for cancer progression, making it an important target in cancer therapy. Lymphangiogenic factors (VEGF-C and VEGF-D) are upregulated in various tumors/cancers, and play an important role in lymphangiogenesis and lymph node metastasis. Similarly, semaphorin 4D (Sema4D) is a potent inducer of angiogenesis, and its overexpression is associated with tumor progression and poor prognosis in a variety of malignancies. However, little is known regarding the functional relationship between Sema4D and VEGF-C/VEGF-D and in the mediation of lymphangiogenesis and lymph node metastasis and clinical outcome. The current study aimed to evaluate the effect of Sema4D expression on outcome in patients with cervical cancer, and to explore the molecular mechanism of Sema4D in tumor progression. We evaluated Sema4D expression, density of lymphatic vessels, and invasion of lymphatic vessels with immunohistochemical methods in 232 human cervical cancers with long-term follow-up. Sema4D expression was correlated with patho-clinical parameters and patients' outcome. A cervical cancer cell line was used to investigate the contribution of sema4D to tumor progression by studying the role of Sema4D in VEGF-C/-D and cell migration using reverse transcription-polymerase chain reaction and Western blotting. We observed that Sema4D expression was higher in metastatic cervical cancer than in nonmetastatic cervical cancer (P<0.001). CD34-positive or D2-40-positive lymphatic vessel density was significantly increased in cases with lymph node metastasis compared with those without lymph node metastasis. The increased Sema4D expression was associated with VEGF-C/-D, the presence of lymphatic invasion, the occurrence of lymph node metastasis, and FIGO stage. We also observed a novel association between Sema4D upregulation and poor prognosis in cervical cancer. In vitro, the Sema4D inhibitory antibody and Sema4D-shRNA suppressed VEGF-C and VEGF-D in the human cervical carcinoma cell lines HeLa, Siha, and Caski cells. Invasiveness assay demonstrated that Sema4D could augment the invasive potential of the tumor cells in the cervical cancer lines and induction of cellular invasiveness by Sema4D stimulation could be inhibited by knockdown of plexinB1 by siRNA. Further mechanistic investigations of tumor cell invasiveness showed that Sema4D could induce activation of GTPase Ras homolog gene family, member A (RhoA), MAPK and AKT. In addition, plexinB1 knockdown by siRNA could suppress the Sema4D signal transmitted to MAPK and Akt. Taken together, these results suggest that Sema4D autocrine within tumor cells contributes to enhanced invasion and tumor progression through increased motility of cervical cancer and VEGF-C/-D-mediated lymphangiogenesis. Sema4D might be useful as a molecular marker of poor prognosis in cervical cancer.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
Sigma-Aldrich
HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
Sigma-Aldrich
HGF mouse, recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture