Merck
  • Consistent and fast inhibition of colon carcinogenesis by polyethylene glycol in mice and rats given various carcinogens.
Consistent and fast inhibition of colon carcinogenesis by polyethylene glycol in mice and rats given various carcinogens.
Cancer research (2000-06-24)
D E Corpet, G Parnaud, M Delverdier, G Peiffer, S Taché
ABSTRACT
We have previously shown that dietary polyethylene-glycol (PEG) suppresses the occurrence of azoxymethane-induced cancers in an accelerated rat model of colon carcinogenesis. To determine the consistency of this preventive effect, we carried out a long-term study in rats fed the standard American Institute of Nutrition 1976 diet, and 7 short-term prevention studies in rodents. A total of 337 F344 rats, 20 Sprague Dawley rats, and 40 OF1 mice were all given initiating dose(s) of colon carcinogen, and were randomly allocated to experimental groups 7 d later. Treated groups received drinking water containing 5% PEG. After 30 or 162 d, the animals were examined for aberrant crypt foci or tumors in the colon. After two 20 mg/kg azoxymethane injections, the number of F344 rats with colon tumor was lower in rats receiving PEG for 162 d than in carcinogen-injected controls, 5/21 versus 25/27 (P < 0.0001). PEG-fed rats had no invasive cancer, and 10 times fewer colon tumors than controls (0.3+/-0.1 and 3.1+/-0.5 respectively, P < 0.0001). A three-day PEG treatment was sufficient to halve the number of azoxymethane-induced aberrant crypt foci in F344 rats (P = 0.0006). After 16 d of treatment, PEG-fed rats had five times fewer foci than controls (21+/-14 and 100+/-23 respectively, P < 0.0001), but the inhibition was reversible in part when treatment was discontinued. Aberrant crypt foci initiated by N-methyl-N-nitrosourea intra-rectally (40 mg/kg) or by 2-amino-3,4-dimethylimidazo(4,5-f)quinoline p.o. (2 x 200 mg/kg) were suppressed by PEG (P < 0.0001 and P = 0.003 respectively). PEG was active in F344 rats, in Sprague Dawley rats (P = 0.0005), and in OF1 mice (P = 0.001). PEGs with MW between 3350 and 12000 (but not PEG 400), and PEG 8000 from five suppliers, markedly inhibited azoxymethane-induced aberrant crypt foci (all P < 0.01). The prevention was stronger in rats fed a high-fat diet (P < 0.0001) than in rats fed a rodent chow (P = 0.02). PEG was thus a fast, consistent, and potent inhibitor of early colonic precursor lesions. Moreover, PEG is one of the most potent inhibitors of colon tumor in the standard rat model. Since PEG has no known toxicity in humans, we think it should be tested as a chemopreventive agent in a clinical trial.
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 8,000, powder
Pricing and availability is not currently available.