Merck
  • Environmental impact of phthalic acid esters and their removal from water and sediments by different technologies--a review.
Environmental impact of phthalic acid esters and their removal from water and sediments by different technologies--a review.
Journal of environmental management (2012-07-17)
Mahmoud M Abdel daiem, José Rivera-Utrilla, Raúl Ocampo-Pérez, José D Méndez-Díaz, Manuel Sánchez-Polo
ABSTRACT
This article describes the most recent methods developed to remove phthalic acid esters (PAEs) from water, wastewater, sludge, and soil. In general, PAEs are considered to be endocrine disrupting chemicals (EDCs), whose effects may not appear until long after exposure. There are numerous methods for removing PAEs from the environment, including physical, chemical and biological treatments, advanced oxidation processes and combinations of these techniques. This review largely focuses on the treatment of PAEs in aqueous solutions but also reports on their treatment in soil and sludge, as well as their effects on human health and the environment.
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Phthalic acid, SAJ special grade, ≥99.0%
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Phthalic acid, analytical standard