Merck
Captopril as a potential inhibitor of lung tumor growth and metastasis.
Annals of the New York Academy of Sciences (2008-10-08)
Samir Attoub, Anne Marie Gaben, Suhail Al-Salam, M A H Al Sultan, Anne John, M Gary Nicholls, Jan Mester, Georg Petroianu
ABSTRACT
Lung cancer is the most common form of cancer in the world, and 90% of patients die from their disease. The angiotensin converting enzyme (ACE) inhibitors are used widely as antihypertensive agents, and it has been suggested that they decrease the risk of some cancers, although available data are conflicting. Accordingly, we investigated the anticancer activity of the ACE inhibitor, captopril, in athymic mice injected with highly tumorigenic LNM35 human lung cells as xenografts. Using this model, we demonstrated that daily IP administration of captopril (2.8 mg/mouse) for 3 weeks resulted in a remarkable reduction of tumor growth (58%, P < 0.01) and lymph node metastasis (50%, P= 0.088). There were no undesirable effects of captopril treatment on animal behavior and body weight. In order to determine the mechanism by which captopril inhibited tumor growth, we investigated the impact of this drug on cell proliferation, apoptosis, and angiogenesis. Immunohistochemical analysis demonstrated that captopril treatment significantly reduced the number of proliferating cells (Ki-67) in the tumor samples but was not associated with inhibition of tumor angiogenesis (CD31). Using cell viability and fluorescent activated cell sorting analysis tests, we demonstrated that captopril inhibited the viability of LNM35 cells by inducing apoptosis, providing insight about the mechanisms underlying its antitumorigenic activities. In view of these experimental findings, we conclude that captopril could be a promising option for the treatment of lung cancer.
Product Description
Captopril for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Captopril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Captopril, meets USP testing specifications
Captopril, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder