Merck
Applications of porous tantalum in total hip arthroplasty.
The Journal of the American Academy of Orthopaedic Surgeons (2006-11-02)
Brett Levine, Craig J Della Valle, Joshua J Jacobs
ABSTRACT
Porous tantalum is an alternative metal for total joint arthroplasty components that offers several unique properties. Its high volumetric porosity (70% to 80%), low modulus of elasticity (3 MPa), and high frictional characteristics make it conducive to biologic fixation. Tantalum has excellent biocompatibility and is safe to use in vivo. The low modulus of elasticity allows for more physiologic load transfer and relative preservation of bone stock. Because of its bioactive nature and ingrowth properties, tantalum is used in primary as well as revision total hip arthroplasty components, with good to excellent early clinical results. In revision arthroplasty, standard and custom augments may serve as a structural bone graft substitute. Formation of a bone-like apatite coating in vivo affords strong fibrous ingrowth properties and allows for substantial soft-tissue attachment, indicating potential for use in cases requiring reattachment of muscles and tendons to a prosthesis. Development of modular components and femoral stems also is being evaluated. The initial clinical data and basic science studies support further investigation of porous tantalum as an alternative to traditional implant materials.
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.9%
Tantalum, tube, 1000mm, outside diameter 3.0mm, inside diameter 2mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, rod, 100mm, diameter 2.0mm, annealed, 99.9%
Tantalum, tube, 100mm, outside diameter 7.0mm, inside diameter 6.7mm, wall thickness 0.15mm, as drawn, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.004mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.034mm, as rolled, 99.9%
Tantalum, tube, 100mm, outside diameter 12.7mm, inside diameter 11.7mm, wall thickness 0.5mm, annealed, 99.9%
Tantalum, tube, 1000mm, outside diameter 3.5mm, inside diameter 3.1mm, wall thickness 0.2mm, as drawn, 99.9%
Tantalum, tube, 100mm, outside diameter 8.7mm, inside diameter 8.34mm, wall thickness 0.18mm, as drawn, 99.9%
Tantalum, tube, 500mm, outside diameter 1.05mm, inside diameter 0.65mm, wall thickness 0.2mm, as drawn, 99.9%
Tantalum, rod, 50mm, diameter 12.7mm, annealed, 99.9%
Tantalum, tube, 200mm, outside diameter 2.0mm, inside diameter 1mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, rod, 25mm, diameter 18mm, annealed, 99.9%
Tantalum, wire reel, 10m, diameter 0.125mm, annealed, 99.9%
Tantalum, wire reel, 2m, diameter 0.8mm, annealed, 99.9%
Tantalum, tube, 200mm, outside diameter 1.02mm, inside diameter 0.86mm, wall thickness 0.08mm, as drawn, 99.9%
Tantalum, foil, 10mm disks, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, 25x25mm, thickness 0.15mm, annealed, 99.9%
Tantalum, foil, 50x50mm, thickness 0.125mm, annealed, 99.9%
Tantalum, foil, 100x100mm, thickness 1.5mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Tantalum, foil, 150x150mm, thickness 0.100mm, annealed, 99.9%
Tantalum, foil, 50x50mm, thickness 0.25mm, annealed, 99.9%
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Tantalum, foil, 0.5m coil, thickness 0.1mm, coil width .62mm, as rolled, 99.9%