Merck
[2Fe-2S] cluster transfer in iron-sulfur protein biogenesis.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-16)
Lucia Banci, Diego Brancaccio, Simone Ciofi-Baffoni, Rebecca Del Conte, Ravisekhar Gadepalli, Maciej Mikolajczyk, Sara Neri, Mario Piccioli, Julia Winkelmann
ABSTRACT
Monothiol glutaredoxins play a crucial role in iron-sulfur (Fe/S) protein biogenesis. Essentially all of them can coordinate a [2Fe-2S] cluster and have been proposed to mediate the transfer of [2Fe-2S] clusters from scaffold proteins to target apo proteins, possibly by acting as cluster transfer proteins. The molecular basis of [2Fe-2S] cluster transfer from monothiol glutaredoxins to target proteins is a fundamental, but still unresolved, aspect to be defined in Fe/S protein biogenesis. In mitochondria monothiol glutaredoxin 5 (GRX5) is involved in the maturation of all cellular Fe/S proteins and participates in cellular iron regulation. Here we show that the structural plasticity of the dimeric state of the [2Fe-2S] bound form of human GRX5 (holo hGRX5) is the crucial factor that allows an efficient cluster transfer to the partner proteins human ISCA1 and ISCA2 by a specific protein-protein recognition mechanism. Holo hGRX5 works as a metallochaperone preventing the [2Fe-2S] cluster to be released in solution in the presence of physiological concentrations of glutathione and forming a transient, cluster-mediated protein-protein intermediate with two physiological protein partners receiving the [2Fe-2S] cluster. The cluster transfer mechanism defined here may extend to other mitochondrial [2Fe-2S] target proteins.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Iron, wire reel, 10m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Iron, wire reel, 20m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 2000m, diameter 0.125mm, hard, 99.5%
Iron, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 100m, diameter 0.065mm, hard, 99.5%
Iron, wire reel, 0.5m, diameter 0.13mm, hard, 99.99+%
Iron, wire reel, 100m, diameter 0.125mm, hard, 99.5%
Iron, wire reel, 1000m, diameter 0.25mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.006mm, 99.85%
Iron, foil, not light tested, 150x150mm, thickness 0.008mm, 99.85%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Iron, foil, 15mm disks, thickness 0.008mm, 99.85%
Iron, foil, 15mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.020mm, 99.85%
Iron, foil, 10mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 25mm disks, thickness 0.003mm, 99.85%
Iron, foil, 1m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 1m coil, thickness 0.025mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.004mm, 99.85%
Iron, foil, 2m coil, thickness 0.38mm, hard, 99.5%