Merck
A new route to C-aryl glycosides.
Organic letters (2002-03-15)
Frank M Hauser, Xingding Hu
ABSTRACT
[reaction: see text] A six-step route for de novo synthesis of C-aryl glycosides based on cycloaddition of an aryl nitrile oxide with 4-pentyn-2-ol has been developed.
Sigma-Aldrich
4-Pentyn-2-ol, ≥98%
Pricing and availability is not currently available.