Merck
  • Polyimide surface modification by using microwave plasma for adhesion enhancement of Cu electroless plating.
Polyimide surface modification by using microwave plasma for adhesion enhancement of Cu electroless plating.
Journal of nanoscience and nanotechnology (2011-07-21)
Sang-Jin Cho, Trieu Nguyen, Jin-Hyo Boo
ABSTRACT
Microwave (MW) plasma was applied to the surface of polyimide (PI) films as a treatment to enhance the adhesion between copper deposition layer and PI surface for electroless plating. The influences of nitrogen MW plasma treatment on chemical composition of the PI surface were investigated by using X-Ray photoelectron spectroscopy (XPS). The wettability was also investigated by water contact angle measurement. The surface morphologies of PI films before and after treatment were characterized with atomic force microscopy (AFM). The contact angle results show that was dramatically decreased to 16.1 degrees at the optimal treatment condition from 72.1 degrees (untreated PI). However, the root mean square (RMS) roughness of treated PI film was almost unchanged. The AFM roughness was stayed from 1.0 to 1.2 with/without plasma treatment. XPS data show a nitrogen increase when PI films exposed to N2 MW plasma. Electroless copper depositions were carried out with the free-formaldehyde method using glyoxylic acid as the reducing reagent and mixture palladium chloride, tin chloride as activation solution. Adhesion property between polyimide surface and copper layer was investigated by tape test.
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O
Pricing and availability is not currently available.