Merck
  • Ingestion of oregano extract increases excretion of urinary phenolic metabolites in humans.
Ingestion of oregano extract increases excretion of urinary phenolic metabolites in humans.
Journal of agricultural and food chemistry (2006-08-31)
Anna Nurmi, Tarja Nurmi, Jaakko Mursu, Raimo Hiltunen, Sari Voutilainen
ABSTRACT
Despite the promising antioxidant action of Lamiaceae herbs in vitro, human studies on these potential sources of dietary antioxidants have remained scarce. In this work, the phenolic acids recovered in human urine after single ingestion of Origanum onites extract were analyzed. The excretion was increased 4- and 2-fold during 0-24 and 24-48 h of the follow-up, respectively. The mean increase in the excretion of phenolic compounds exceeded the ingested amount of identified phenolic acids. The result can be partly explained by rosmarinic acid, the main identified phenolic constituent in the extract, as well as flavonoids present in minor amounts, presumably being metabolized into a double amount of simple phenolic metabolites. Furthermore, unidentified phenolic constituents in the extract partly contribute to the excretory increase. The main metabolite, p-hydroxybenzoic acid, was excreted rapidly. The results show that constituents of oregano extract and, in particular, their metabolites may contribute to the dietary intake of phenolic antioxidants.
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Pricing and availability is not currently available.