Merck
  • A novel series of (S)-2,7-substituted-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acids: peroxisome proliferator-activated receptor α/γ dual agonists with protein-tyrosine phosphatase 1B inhibitory activity.
A novel series of (S)-2,7-substituted-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acids: peroxisome proliferator-activated receptor α/γ dual agonists with protein-tyrosine phosphatase 1B inhibitory activity.
Chemical & pharmaceutical bulletin (2011-10-04)
Kazuya Otake, Satoru Azukizawa, Masaki Fukui, Michiko Shibabayashi, Hikaru Kamemoto, Tomohiro Miike, Kazuyoshi Kunishiro, Masayasu Kasai, Hiroaki Shirahase
ABSTRACT
Novel 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid derivatives were synthesized and (S)-7-(2-{2-[(E)-2-cyclopentylvinyl]-5-methyloxazol-4-yl}ethoxy)-2-[(2E,4E)-hexadienoyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (14c) was identified as a peroxisome proliferator-activated receptor (PPAR) α/γ dual agonist. The transactivation activity of 14c was comparable to that of rosiglitazone in human PPARγ (EC50=0.14 µM) and was much higher than in human PPARα (EC50=0.20 µM). In addition, 14c, but not rosiglitazone, showed human protein-tyrosine phosphatase 1B (PTP-1B) inhibitory activity (IC50=1.85 µM). 14c showed about 10-fold stronger hypoglycemic and hypotriglyceridemic effects than rosiglitazone by repeated application for 14 d in male KK-Ay mice. Furthermore, 14c, but not rosiglitazone, increased hepatic peroxisome acyl CoA oxidase activity at 30 mg/kg/d for 7 d in male Syrian hamsters, probably due to its PPARα agonist activity. 14c did not affect plasma volume at 100 mg/kg/d for 14 d in male ICR mice, while rosiglitazone significantly increased it. In conclusion, 14c is a promising candidate for an efficacious and safe anti-diabetic drug with triple actions as a PPARα/γ dual agonist with PTP-1B inhibitory activity.
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%
Pricing and availability is not currently available.